Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029402.3 | NP_083678.2 | 14 - 15 | 1586 | 89 | |
GenBank mRNA | AK004875.2 | - | 14 - 15 | 1542 | 89 | |
AK016520.1 | - | 14 - 15 | 1556 | 89 | ||
AK028589.1 | - | 14 - 15 | 1555 | 89 | ||
AK144693.1 | - | 14 - 15 | 1560 | 89 | ||
AK160597.1 | - | 14 - 15 | 1571 | 89 | ||
AK220287.1 | - | 14 - 15 | 1561 | 89 | ||
BC025902.1 | - | 6 - 7 | 652 | 89 | ||
BC026779.1 | - | 14 - 15 | 1487 | 4018298 | 89 | |
BC027428.1 | - | 14 - 15 | 1549 | 3983461 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |