Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029402.3 | NP_083678.2 | 15 - 16 | 1733 | 92 | |
GenBank mRNA | AK004875.2 | - | 15 - 16 | 1689 | 92 | |
AK016520.1 | - | 15 - 16 | 1703 | 92 | ||
AK028589.1 | - | 15 - 16 | 1702 | 92 | ||
AK144693.1 | - | 15 - 16 | 1707 | 92 | ||
AK160597.1 | - | 15 - 16 | 1718 | 92 | ||
AK220287.1 | - | 15 - 16 | 1708 | 92 | ||
BC025902.1 | - | 7 - 8 | 799 | 92 | ||
BC026779.1 | - | 15 - 16 | 1634 | 4018298 | 92 | |
BC027428.1 | - | 15 - 16 | 1696 | 3983461 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |