Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030113.2 | NP_084389.2 | 6 - 7 | 723 | 95 | |
GenBank mRNA | AF297029.1 | - | 6 - 7 | 627 | 95 | |
AF297030.1 | - | 6 - 7 | 686 | 95 | ||
AY179965.1 | - | 6 - 7 | 599 | 95 | ||
AY179966.1 | - | 6 - 7 | 599 | 95 | ||
AY179967.1 | - | 6 - 7 | 599 | 95 | ||
CV783737.1 | - | 1 - 2 | 101 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |