Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030132.5 | NP_084408.1 | 1 - 2 | 274 | 112 | |
GenBank mRNA | AK021306.1 | - | 1 - 2 | 224 | 112 | |
AK134445.1 | - | 1 - 2 | 206 | 112 | ||
BC046791.1 | - | 1 - 2 | 219 | 5717427 | 112 | |
BC056961.1 | - | 1 - 2 | 219 | 5717427 | 112 | |
CK334116.1 | - | 1 - 2 | 286 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |