Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031402.2 | NP_113579.2 | 4 - 5 | 926 | 64 | |
GenBank mRNA | AF329198.1 | - | 4 - 5 | 923 | 64 | |
AK132405.1 | - | 4 - 5 | 664 | 64 | ||
AK133870.1 | - | 4 - 5 | 927 | 64 | ||
AK134273.1 | - | 4 - 5 | 862 | 64 | ||
AK137611.1 | - | 4 - 5 | 927 | 64 | ||
BC040768.1 | - | 4 - 5 | 1255 | 4458200 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |