Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130457.2 | NP_569724.2 | 22 - 23 | 3759 | 61 | |
GenBank mRNA | AF333770.1 | - | 22 - 23 | 3724 | 61 | |
AK053611.1 | - | 20 - 21 | 3361 | 61 | ||
AK142816.1 | - | 22 - 23 | 3755 | 61 | ||
AK173255.1 | - | 22 - 23 | 3734 | 61 | ||
BC034628.1 | - | 22 - 23 | 3759 | 5369648 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |