Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133798.3 | NP_598559.2 | 2 - 3 | 649 | 129 | |
GenBank mRNA | AK146901.1 | - | 2 - 3 | 530 | 129 | |
AK147922.1 | - | 2 - 3 | 509 | 129 | ||
AK158201.1 | - | 2 - 3 | 540 | 129 | ||
AK162206.1 | - | 1 - 2 | 417 | 129 | ||
AK164329.1 | - | 2 - 3 | 525 | 129 | ||
AK166390.1 | - | 2 - 3 | 531 | 129 | ||
AK167692.1 | - | 2 - 3 | 650 | 129 | ||
BC018508.1 | - | 1 - 2 | 192 | 3156447 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |