Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025365.2 | NP_001020536.2 | 2 - 3 | 539 | 67 | |
GenBank mRNA | AK133976.1 | - | 4 - 5 | 691 | 67 | |
AK170224.1 | - | 2 - 3 | 332 | 67 | ||
BC017525.1 | - | 2 - 3 | 282 | 4910951 | 67 | |
BC024069.1 | - | 2 - 3 | 321 | 5097088 | 67 | |
FJ618908.1 | - | 2 - 3 | 539 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |