Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133930.1 | NP_598691.1 | 6 - 7 | 1060 | 58 | |
GenBank mRNA | AB009020.1 | - | 6 - 7 | 1065 | 58 | |
AK077726.1 | - | 6 - 7 | 1205 | 58 | ||
AK155777.1 | - | 6 - 7 | 1070 | 58 | ||
BC010804.1 | - | 6 - 7 | 1060 | 4239786 | 58 | |
BC023893.1 | - | 6 - 7 | 1071 | 5310111 | 58 | |
BC025932.1 | - | 6 - 7 | 966 | 5102571 | 58 | |
BC029065.1 | - | 6 - 7 | 1096 | 4935296 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |