Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286030.1 | NP_001272959.1 | 2 - 3 | 756 | 95 | |
NM_001286031.1 | NP_001272960.1 | 2 - 3 | 625 | 95 | ||
NM_133941.2 | NP_598702.1 | 2 - 3 | 709 | 95 | ||
GenBank mRNA | AK028910.1 | - | 2 - 3 | 566 | 95 | |
AK033624.1 | - | 2 - 3 | 711 | 95 | ||
AK167889.1 | - | 2 - 3 | 654 | 95 | ||
AK169005.1 | - | 2 - 3 | 700 | 95 | ||
AK172504.1 | - | 2 - 3 | 690 | 95 | ||
AK204739.1 | - | 1 - 2 | 284 | 95 | ||
AY065980.1 | - | 2 - 3 | 711 | 95 | ||
BC006911.1 | - | 1 - 2 | 266 | 95 | ||
BC022920.1 | - | 1 - 2 | 465 | 4460064 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |