Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253847.1 | NP_001240776.1 | 3 - 4 | 363 | 119 | |
NM_001253848.1 | NP_001240777.1 | 3 - 4 | 351 | 119 | ||
NM_134048.2 | NP_598809.1 | 3 - 4 | 360 | 119 | ||
GenBank mRNA | AF167441.1 | - | 3 - 4 | 363 | 119 | |
AK049141.1 | - | 3 - 4 | 299 | 119 | ||
AK051737.1 | - | 3 - 4 | 302 | 119 | ||
AK166017.1 | - | 3 - 4 | 293 | 119 | ||
BB651673.1 | - | 3 - 4 | 294 | 119 | ||
BC019529.1 | - | 3 - 4 | 284 | 4219836 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |