Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291039.1 | NP_001277968.1 | 8 - 9 | 726 | 100 | |
NM_138651.7 | NP_619592.1 | 8 - 9 | 1049 | 100 | ||
GenBank mRNA | AK036328.1 | - | 8 - 9 | 900 | 100 | |
AK049257.1 | - | 8 - 9 | 727 | 100 | ||
AK147541.1 | - | 8 - 9 | 941 | 100 | ||
AK166001.1 | - | 8 - 9 | 900 | 100 | ||
AY159802.1 | - | 8 - 9 | 972 | 100 | ||
BC003852.1 | - | 8 - 9 | 955 | 3600825 | 100 | |
BC069879.1 | - | 8 - 9 | 928 | 30439069 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |