Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253768.1 | NP_001240697.1 | 4 - 5 | 747 | 74 | |
NM_001253769.1 | NP_001240698.1 | 3 - 4 | 643 | 74 | ||
NM_001253770.1 | NP_001240699.1 | 3 - 4 | 492 | 74 | ||
NM_138744.3 | NP_620083.1 | 3 - 4 | 492 | 74 | ||
GenBank mRNA | AF532969.1 | - | 2 - 3 | 287 | 74 | |
AK031356.1 | - | 4 - 5 | 471 | 74 | ||
AK043865.1 | - | 3 - 4 | 596 | 74 | ||
AK049080.1 | - | 3 - 4 | 644 | 74 | ||
AK077112.1 | - | 4 - 5 | 532 | 74 | ||
AK173064.1 | - | 3 - 4 | 479 | 74 | ||
BC021749.1 | - | 3 - 4 | 447 | 4165430 | 74 | |
BC031527.1 | - | 3 - 4 | 455 | 4008624 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |