Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139309.4 | NP_647470.1 | 3 - 4 | 480 | 68 | |
GenBank mRNA | AB077383.1 | - | 4 - 5 | 664 | 68 | |
AJ511807.1 | - | 4 - 5 | 500 | 68 | ||
AK052357.1 | - | 5 - 6 | 638 | 68 | ||
AK085931.1 | - | 3 - 4 | 481 | 68 | ||
AK142744.1 | - | 640 | 68 | |||
AK144743.1 | - | 4 - 5 | 509 | 68 | ||
BC017538.1 | - | 4 - 5 | 496 | 4506804 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |