Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160215.1 | NP_001153687.1 | 3 - 4 | 572 | 73 | |
NM_145141.2 | NP_660123.1 | 3 - 4 | 569 | 73 | ||
NR_027666.1 | 3 - 4 | 638 | 73 | |||
GenBank mRNA | AF329487.1 | - | 3 - 4 | 536 | 73 | |
AF426462.1 | - | 3 - 4 | 529 | 73 | ||
AF531424.1 | - | 3 - 4 | 558 | 73 | ||
AK041572.1 | - | 3 - 4 | 549 | 73 | ||
AK172370.1 | - | 3 - 4 | 562 | 73 | ||
BC064708.1 | - | 3 - 4 | 572 | 30301438 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |