Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145365.3 | NP_663340.1 | 10 - 11 | 1379 | 86 | |
GenBank mRNA | AF392874.1 | - | 8 - 9 | 1030 | 86 | |
AK050186.1 | - | 8 - 9 | 1021 | 86 | ||
AK077258.1 | - | 10 - 11 | 1380 | 86 | ||
BC010786.1 | - | 8 - 9 | 1074 | 4211480 | 86 | |
BC028820.1 | - | 8 - 9 | 1051 | 5096773 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |