Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145373.2 | NP_663348.2 | 4 - 5 | 575 | 70 | |
GenBank mRNA | AK082020.1 | - | 3 - 4 | 267 | 70 | |
BC010462.1 | - | 4 - 5 | 542 | 3585040 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |