Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351036 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351038 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351039 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351037 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2078073 |
BC019216.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2078073 |
BC025609.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2078075 |
BC019216.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355219 |
NM_009603.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355219 |
X55718.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
749251 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667799 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
550268 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
140090 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455152 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
970611 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
673243 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839459 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545378 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
36331 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
147133 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P119697 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P130369 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2078073 |
BC019216.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2078073 |
BC025609.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2078075 |
BC019216.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351009 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351034 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351032 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351010 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351005 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351026 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351024 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351019 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351036 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351016 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351039 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351011 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351015 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351012 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351037 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351023 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351018 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351028 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351003 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351030 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351035 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351014 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351004 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23351027 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05424683 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P119697 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01003404 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05424703 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P130369 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P306892 |
BC019216.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306892 |
BC025609.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130200 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P119697 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P130369 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03846673 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03481829 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03846674 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03481816 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03481803 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351037 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351039 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351036 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23351038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23350948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23351019 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351006 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350988 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351032 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04050676 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350987 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351019 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351039 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350983 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04080450 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351011 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351009 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351000 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350982 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350973 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351024 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03481803 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04050678 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04050680 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350976 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351028 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351014 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350995 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351010 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350986 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351034 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03205223 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350997 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351018 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350981 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351023 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351005 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351004 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351027 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351016 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23350999 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351002 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351031 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03481816 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351013 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351030 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23351001 |
- |
- |