Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130194.1 | NP_001123666.1 | 7 - 8 | 954 | 111 | |
NR_052011.1 | 7 - 8 | 1195 | 111 | |||
GenBank mRNA | AY450428.1 | - | 7 - 8 | 1196 | 111 | |
BC019528.1 | - | 6 - 7 | 928 | 4208860 | 111 | |
BC031186.1 | - | 6 - 7 | 915 | 4989959 | 111 | |
BC036157.1 | - | 7 - 8 | 954 | 4982413 | 111 | |
BC036163.1 | - | 7 - 8 | 995 | 4984410 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |