Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141976 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141979 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141974 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141977 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141981 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141978 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967880 |
BC023107.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967880 |
AK165622.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967880 |
AK165993.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967880 |
AK088257.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967880 |
NM_145412.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396081 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
523755 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
369868 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
30105 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
331243 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577736 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
929424 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
653626 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837010 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
970229 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
79777 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853451 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
27730 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647544 |
AF194972.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647544 |
AK031299.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647544 |
AK041821.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647544 |
AK049279.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647544 |
AK220327.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647544 |
NM_175472.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106349 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
98598 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81251 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
73943 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P137193 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P104328 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967880 |
BC023107.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967880 |
AK165622.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967880 |
AK165993.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967880 |
AK088257.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967880 |
NM_145412.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141953 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141969 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141988 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141965 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141974 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141967 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141950 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141979 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141939 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141962 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141961 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141992 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142018 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142005 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141982 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141973 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141944 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141955 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141981 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142024 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141987 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141947 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141942 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141984 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141994 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141996 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142021 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141971 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141952 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141948 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142007 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141990 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141991 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141977 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141998 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141963 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142022 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142020 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142010 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141968 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142003 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142012 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141949 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142006 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142001 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142008 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141957 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142016 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141959 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26142013 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141972 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26141941 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01820365 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P104328 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P171557 |
BC023107.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P171557 |
AK194778.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P171557 |
AK165622.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P171557 |
AK088257.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P171557 |
NM_145412.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P420541 |
AK087721.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109040 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P104328 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07095957 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06724613 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06913521 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07295594 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06724610 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07295590 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06913520 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07355926 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06987840 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06987845 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141981 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141976 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141978 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141974 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141979 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26141977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141901 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141918 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141840 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141872 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26141845 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06928516 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141967 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141918 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141909 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07369669 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141968 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141957 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141929 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141969 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141944 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06928508 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141947 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141931 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141960 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141924 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141935 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141900 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141955 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141936 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141901 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141921 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07279449 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141965 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141905 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141942 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141973 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141938 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141963 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141953 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141917 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141972 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141956 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141974 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26141940 |
- |
- |