Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145416.3 | NP_663391.2 | 2 - 3 | 184 | 106 | |
GenBank mRNA | AK051997.1 | - | 2 - 3 | 185 | 106 | |
BC021438.1 | - | 2 - 3 | 177 | 5039543 | 106 | |
CJ069606.1 | - | 2 - 3 | 186 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |