Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145425.3 | NP_663400.2 | 14 - 15 | 1936 | 79 | |
GenBank mRNA | AF424701.1 | - | 14 - 15 | 1899 | 79 | |
AK043672.1 | - | 13 - 14 | 1823 | 79 | ||
AK083042.1 | - | 14 - 15 | 1937 | 79 | ||
BC141166.1 | - | 14 - 15 | 1900 | 9055778 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |