Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145431.2 | NP_663406.2 | 8 - 9 | 1004 | 62 | |
GenBank mRNA | AK141230.1 | - | 8 - 9 | 979 | 62 | |
AK170853.1 | - | 8 - 9 | 977 | 62 | ||
BC018399.1 | - | 8 - 9 | 995 | 3491925 | 62 | |
BC035421.1 | - | 8 - 9 | 995 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |