Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Llgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252532.1 | NP_001239461.1 | 14 - 15 | 1733 | 77 | |
NM_145438.2 | NP_663413.2 | 14 - 15 | 1733 | 77 | ||
GenBank mRNA | AK143486.1 | - | 14 - 15 | 1720 | 77 | |
AK143798.1 | - | 14 - 15 | 1707 | 77 | ||
AK167536.1 | - | 14 - 15 | 1705 | 77 | ||
AY033650.1 | - | 14 - 15 | 1683 | 77 | ||
BC013232.1 | - | 10 - 11 | 1274 | 77 | ||
BC037226.1 | - | 14 - 15 | 1705 | 3584689 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |