Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145471.2 | NP_663446.1 | 4 - 5 | 681 | 77 | |
GenBank mRNA | AK036278.1 | - | 2 - 3 | 554 | 77 | |
AK132158.1 | - | 2 - 3 | 1245 | 77 | ||
AK149655.1 | - | 2 - 3 | 643 | 77 | ||
AK159836.1 | - | 2 - 3 | 624 | 77 | ||
AY239226.1 | - | 4 - 5 | 594 | 77 | ||
BC022126.1 | - | 4 - 5 | 714 | 5097371 | 77 | |
BC138217.1 | - | 2 - 3 | 855 | 8861236 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |