Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145515.2 | NP_663490.2 | 9 - 10 | 985 | 98 | |
GenBank mRNA | AB185282.1 | - | 9 - 10 | 908 | 98 | |
AF453686.1 | - | 9 - 10 | 915 | 98 | ||
AK173181.1 | - | 8 - 9 | 835 | 98 | ||
BC112400.1 | - | 9 - 10 | 914 | 40059725 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |