Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145528.3 | NP_663503.1 | 13 - 14 | 1479 | 79 | |
GenBank mRNA | AK122339.1 | - | 9 - 10 | 3841 | 79 | |
AK149672.1 | - | 13 - 14 | 1438 | 79 | ||
AK154470.1 | - | 13 - 14 | 1467 | 79 | ||
AK163962.1 | - | 13 - 14 | 1433 | 79 | ||
AK171206.1 | - | 13 - 14 | 1465 | 79 | ||
AK171440.1 | - | 13 - 14 | 1456 | 79 | ||
BC016669.1 | - | 13 - 14 | 1463 | 2648797 | 79 | |
BC023673.1 | - | 13 - 14 | 1429 | 5343236 | 79 | |
BC023712.1 | - | 13 - 14 | 1452 | 5346303 | 79 | |
BC033419.1 | - | 13 - 14 | 1430 | 5359944 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |