Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146046.1 | NP_001139518.1 | 7 - 8 | 983 | 63 | |
NM_001146047.1 | NP_001139519.1 | 8 - 9 | 982 | 63 | ||
NM_145616.4 | NP_663591.3 | 8 - 9 | 1181 | 63 | ||
GenBank mRNA | AK041627.1 | - | 6 - 7 | 685 | 63 | |
AK049268.1 | - | 8 - 9 | 960 | 63 | ||
AK051325.1 | - | 7 - 8 | 788 | 63 | ||
AK085022.1 | - | 976 | 63 | |||
BC016574.1 | - | 7 - 8 | 750 | 3594577 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |