Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145925.2 | NP_666037.1 | 1 - 2 | 160 | 67 | |
GenBank mRNA | AK035696.1 | - | 1 - 2 | 175 | 67 | |
AK075773.1 | - | 1 - 2 | 133 | 67 | ||
AK088813.1 | - | 1 - 2 | 197 | 67 | ||
AK089887.1 | - | 1 - 2 | 142 | 67 | ||
AK098093.1 | - | 1 - 2 | 174 | 67 | ||
AK151639.1 | - | 1 - 2 | 142 | 67 | ||
AK152366.1 | - | 1 - 2 | 142 | 67 | ||
AK159987.1 | - | 1 - 2 | 142 | 67 | ||
AK167972.1 | - | 1 - 2 | 172 | 67 | ||
AK168164.1 | - | 1 - 2 | 172 | 67 | ||
AK170802.1 | - | 1 - 2 | 142 | 67 | ||
AK170971.1 | - | 1 - 2 | 175 | 67 | ||
AK170972.1 | - | 1 - 2 | 142 | 67 | ||
AK171140.1 | - | 1 - 2 | 141 | 67 | ||
BC023961.1 | - | 1 - 2 | 215 | 5323397 | 67 | |
BC025533.1 | - | 1 - 2 | 184 | 5250502 | 67 | |
BC029144.1 | - | 1 - 2 | 156 | 4945974 | 67 | |
BC032184.1 | - | 1 - 2 | 179 | 5310191 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |