Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269021 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269020 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269025 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269022 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269023 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC032184.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC023961.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC025533.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC029144.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
AK202715.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC032184.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC023961.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC025533.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
BC029144.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P173100 |
AK202715.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439247 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359974 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
827695 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
622384 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
283276 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
128161 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
242426 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479187 |
AK015992.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479187 |
AK016431.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479187 |
AK030125.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479187 |
BC132158.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479187 |
NM_028737.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
691209 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
419330 |
AK047306.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
91689 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
119861 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P163443 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144665 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC032184.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC023961.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC025533.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC029144.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
AK202715.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC032184.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC023961.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC025533.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
BC029144.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P173100 |
AK202715.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269008 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269044 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269041 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269024 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269039 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269033 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269020 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269032 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269021 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269035 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269025 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269012 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269022 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269013 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269016 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269030 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269015 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269018 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269027 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269009 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27269028 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07115879 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P163443 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P144665 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC032184.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC023961.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC025533.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC029144.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
AK202715.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC032184.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC023961.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC025533.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
BC029144.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P173100 |
AK202715.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P107282 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P163443 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P144665 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07115879 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08936134 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08740527 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08936121 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08936132 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08740523 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269025 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269021 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269022 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269023 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27269020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27268996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27269027 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08936121 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269065 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269010 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08542556 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269059 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269073 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269057 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08690777 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08266169 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08458608 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269067 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269004 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269058 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08633923 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269012 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269063 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08740528 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269044 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269041 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08936119 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08633902 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08210380 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08633904 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269051 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269020 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269015 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27268997 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269035 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269049 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269056 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269039 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269066 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269026 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269046 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269032 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269033 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269025 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269002 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269007 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269038 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08740524 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269060 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269000 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269030 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08695392 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08690779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27269022 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08633921 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08431741 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08961023 |
- |
- |