Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145926.3 | NP_666038.3 | 8 - 9 | 1217 | 62 | |
GenBank mRNA | AB053218.1 | - | 8 - 9 | 919 | 62 | |
AK177701.1 | - | 3 - 4 | 231 | 62 | ||
AK190141.1 | - | 231 | 62 | |||
AK191887.1 | - | 3 - 4 | 231 | 62 | ||
AK192181.1 | - | 3 - 4 | 231 | 62 | ||
AK195601.1 | - | 3 - 4 | 231 | 62 | ||
AK199018.1 | - | 3 - 4 | 231 | 62 | ||
AK204039.1 | - | 3 - 4 | 231 | 62 | ||
AK205336.1 | - | 3 - 4 | 63 | |||
AK213265.1 | - | 3 - 4 | 231 | 62 | ||
AK217144.1 | - | 3 - 4 | 231 | 62 | ||
BC026638.1 | - | 3 - 4 | 325 | 4984907 | 62 | |
BC031613.1 | - | 2 - 3 | 195 | 4216720 | 62 | |
CX205743.1 | - | 5 - 6 | 477 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |