Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145928.1 | NP_666040.1 | 4 - 5 | 322 | 67 | |
GenBank mRNA | AK030593.1 | - | 4 - 5 | 324 | 67 | |
AK156671.1 | - | 4 - 5 | 334 | 67 | ||
BC024611.1 | - | 4 - 5 | 327 | 4188261 | 67 | |
BC025568.1 | - | 4 - 5 | 336 | 5322531 | 67 | |
BC026574.1 | - | 4 - 5 | 322 | 4954874 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |