Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145928.1 | NP_666040.1 | 6 - 7 | 623 | 70 | |
GenBank mRNA | AK030593.1 | - | 6 - 7 | 625 | 70 | |
AK156671.1 | - | 6 - 7 | 636 | 70 | ||
BC024611.1 | - | 6 - 7 | 628 | 4188261 | 70 | |
BC025568.1 | - | 6 - 7 | 637 | 5322531 | 70 | |
BC026574.1 | - | 6 - 7 | 623 | 4954874 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |