Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145932.3 | NP_666044.1 | 3 - 4 | 649 | 76 | |
GenBank mRNA | AK052725.1 | - | 3 - 4 | 650 | 76 | |
AK147155.1 | - | 469 | 76 | |||
BC024441.1 | - | 3 - 4 | 457 | 4973857 | 76 | |
BC025912.1 | - | 3 - 4 | 460 | 5064489 | 76 | |
BC031178.1 | - | 3 - 4 | 400 | 4988213 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |