Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148935.2 | NP_683737.2 | 1 - 2 | 74 | 58 | |
GenBank mRNA | AF323488.1 | - | 1 - 2 | 82 | 58 | |
AY039039.1 | - | 1 - 2 | 120 | 58 | ||
BC092242.1 | - | 1 - 2 | 63 | 30698162 | 58 | |
BC130222.1 | - | 1 - 2 | 75 | 40141600 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |