Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479025 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479023 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479022 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479021 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2098688 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1972852 |
AK215222.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1972852 |
AK217746.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1972852 |
BC003746.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1972852 |
BC085099.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1972852 |
BC108340.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
944987 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
525462 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
AK136052.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
AK136732.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
AK138724.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
AK172036.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
BC096553.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
NM_011083.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
U52193.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577672 |
U55772.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
21622 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169258 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1972852 |
AK215222.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1972852 |
AK217746.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1972852 |
BC003746.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1972852 |
BC085099.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1972852 |
BC108340.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2098688 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479031 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479004 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479009 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479024 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479010 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479025 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479028 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479003 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479007 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479033 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479037 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479023 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479013 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30479036 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169258 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08478397 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P107126 |
AF015309.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P682450 |
BC003746.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P682450 |
BC085099.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P682450 |
BC108340.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P682450 |
AK167242.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P682450 |
AK158611.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117123 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169258 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12003271 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12003277 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12003266 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12003267 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12003274 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479021 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479023 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479025 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30479022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478901 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478894 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478892 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478885 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30479028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30478980 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479031 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479028 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478991 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479000 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479008 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479004 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479042 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478986 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478980 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478954 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12161397 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478999 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11959772 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479010 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478995 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12445962 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478956 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479025 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479003 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479032 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479021 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12185002 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478970 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478959 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478958 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478969 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478982 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478983 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478961 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479045 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12445961 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478998 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478976 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478953 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11978141 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12370164 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12003274 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478993 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12445964 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478960 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478965 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478962 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478968 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478979 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478985 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30478981 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30479039 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12394559 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12394554 |
- |
- |