Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019694.1 | NP_062668.1 | 12 - 13 | 2076 | 95 | |
GenBank mRNA | AB041582.1 | - | 12 - 13 | 2089 | 95 | |
AF061026.1 | - | 12 - 13 | 2076 | 95 | ||
AK133849.1 | - | 12 - 13 | 2096 | 95 | ||
AK161638.1 | - | 5 - 6 | 2835 | 95 | ||
AK207004.1 | - | 2 - 3 | 203 | 95 | ||
AK210262.1 | - | 1 - 2 | 130 | 95 | ||
AK215386.1 | - | 1 - 2 | 157 | 95 | ||
AK217209.1 | - | 1 - 2 | 128 | 95 | ||
BC023862.1 | - | 12 - 13 | 2073 | 95 | ||
BC046326.1 | - | 12 - 13 | 2083 | 95 | ||
BC055865.1 | - | 12 - 13 | 2094 | 95 | ||
BC061690.1 | - | 12 - 13 | 2075 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |