Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144786.2 | NP_659035.2 | 6 - 7 | 868 | 82 | |
GenBank mRNA | AK032051.1 | - | 6 - 7 | 866 | 82 | |
BC005772.1 | - | 6 - 7 | 849 | 3602314 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |