Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144813.1 | NP_659062.1 | 6 - 7 | 2390 | 62 | |
GenBank mRNA | BC016094.1 | - | 6 - 7 | 2390 | 4504496 | 62 |
BC044859.1 | - | 5 - 6 | 1826 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |