Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cecr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144815.2 | NP_659064.1 | 2 - 3 | 336 | 70 | |
GenBank mRNA | AK044228.1 | - | 2 - 3 | 536 | 70 | |
AK140387.1 | - | 2 - 3 | 357 | 70 | ||
BC014705.1 | - | 2 - 3 | 345 | 4238201 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |