Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144822.3 | NP_659071.1 | 4 - 5 | 627 | 87 | |
GenBank mRNA | AK049521.1 | - | 4 - 5 | 623 | 87 | |
AK050113.1 | - | 4 - 5 | 614 | 87 | ||
AK052485.1 | - | 4 - 5 | 575 | 87 | ||
AK077605.1 | - | 4 - 5 | 577 | 87 | ||
AK085553.1 | - | 4 - 5 | 546 | 87 | ||
AK189367.1 | - | 1 - 2 | 50 | 87 | ||
AK200391.1 | - | 1 - 2 | 50 | 87 | ||
BC018320.1 | - | 4 - 5 | 568 | 4192966 | 87 | |
BC023042.1 | - | 3 - 4 | 511 | 5357794 | 87 | |
BC026566.1 | - | 4 - 5 | 578 | 4977083 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |