Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144865.2 | NP_659114.2 | 5 - 6 | 587 | 80 | |
GenBank mRNA | AY562230.1 | - | 5 - 6 | 420 | 80 | |
BC020184.1 | - | 5 - 6 | 518 | 4505679 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |