Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144866.3 | NP_659115.3 | 5 - 6 | 727 | 75 | |
GenBank mRNA | AK049607.1 | - | 5 - 6 | 678 | 75 | |
AK139933.1 | - | 5 - 6 | 690 | 75 | ||
AK151028.1 | - | 5 - 6 | 628 | 75 | ||
AK151710.1 | - | 5 - 6 | 628 | 75 | ||
AK152428.1 | - | 5 - 6 | 628 | 75 | ||
AK152952.1 | - | 5 - 6 | 628 | 75 | ||
AK164020.1 | - | 5 - 6 | 678 | 75 | ||
AK166740.1 | - | 5 - 6 | 696 | 75 | ||
AK169345.1 | - | 5 - 6 | 700 | 75 | ||
AK170221.1 | - | 5 - 6 | 670 | 75 | ||
BC013717.1 | - | 5 - 6 | 654 | 3992883 | 75 | |
D87691.1 | - | 2 - 3 | 242 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |