Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039482.1 | NP_001034571.1 | 4 - 5 | 915 | 55 | |
GenBank mRNA | AK032131.1 | - | 2 - 3 | 402 | 55 | |
AK053102.1 | - | 4 - 5 | 865 | 55 | ||
AK054169.1 | - | 4 - 5 | 883 | 55 | ||
AK220547.1 | - | 4 - 5 | 894 | 55 | ||
BC019571.1 | - | 4 - 5 | 887 | 4235202 | 55 | |
BC060183.1 | - | 4 - 5 | 844 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |