Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039482.1 | NP_001034571.1 | 5 - 6 | 1006 | 91 | |
GenBank mRNA | AK032131.1 | - | 3 - 4 | 493 | 91 | |
AK053102.1 | - | 5 - 6 | 956 | 91 | ||
AK054169.1 | - | 5 - 6 | 974 | 91 | ||
AK220547.1 | - | 5 - 6 | 985 | 91 | ||
BC019571.1 | - | 5 - 6 | 978 | 4235202 | 91 | |
BC060183.1 | - | 5 - 6 | 935 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |