Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144879.2 | NP_659128.2 | 5 - 6 | 917 | 72 | |
NR_027352.1 | 5 - 6 | 784 | 72 | |||
GenBank mRNA | AK045261.1 | - | 5 - 6 | 713 | 72 | |
AK078596.1 | - | 5 - 6 | 786 | 72 | ||
AK140989.1 | - | 5 - 6 | 911 | 72 | ||
AK163193.1 | - | 5 - 6 | 693 | 72 | ||
BC024141.1 | - | 5 - 6 | 687 | 5149750 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |