Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100031293.mm.1 |
NM_144881.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031293.mm.1 |
NM_144881.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031293.mm.1 |
TC0100003827.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031295.mm.1 |
NM_144881.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031295.mm.1 |
NM_144881.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031295.mm.1 |
TC0100003827.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031292.mm.1 |
NM_144881.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031292.mm.1 |
NM_144881.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031292.mm.1 |
TC0100003827.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031294.mm.1 |
NM_144881.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031294.mm.1 |
NM_144881.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031294.mm.1 |
TC0100003827.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031296.mm.1 |
NM_144881.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031296.mm.1 |
NM_144881.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031296.mm.1 |
TC0100003827.mm.1 |
MTA TC |