Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144903.2 | NP_659152.1 | 2 - 3 | 171 | 60 | |
GenBank mRNA | AK135385.1 | - | 2 - 3 | 200 | 60 | |
AK149395.1 | - | 2 - 3 | 188 | 60 | ||
AK149598.1 | - | 2 - 3 | 185 | 60 | ||
AK167334.1 | - | 2 - 3 | 188 | 60 | ||
AK167566.1 | - | 2 - 3 | 185 | 60 | ||
AK168600.1 | - | 2 - 3 | 185 | 60 | ||
AK168997.1 | - | 2 - 3 | 235 | 60 | ||
BC016435.1 | - | 2 - 3 | 167 | 4241025 | 60 | |
BC022113.1 | - | 2 - 3 | 144 | 5098524 | 60 | |
BC024056.1 | - | 2 - 3 | 177 | 5100067 | 60 | |
BC024112.1 | - | 2 - 3 | 176 | 5102118 | 60 | |
BC026577.1 | - | 2 - 3 | 136 | 4989521 | 60 | |
BC030724.1 | - | 2 - 3 | 173 | 5100784 | 60 | |
BC030725.1 | - | 2 - 3 | 177 | 5101686 | 60 | |
BC034169.1 | - | 2 - 3 | 177 | 4975382 | 60 | |
BC034171.1 | - | 2 - 3 | 154 | 4972521 | 60 | |
BC034172.1 | - | 2 - 3 | 141 | 4974489 | 60 | |
BC034173.1 | - | 2 - 3 | 153 | 4974553 | 60 | |
BC036130.1 | - | 2 - 3 | 154 | 4974071 | 60 | |
BC036131.1 | - | 2 - 3 | 171 | 4974877 | 60 | |
BC036132.1 | - | 2 - 3 | 171 | 4975336 | 60 | |
BC036133.1 | - | 2 - 3 | 183 | 4975573 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |