Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gckr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144909.1 | NP_659158.1 | 2 - 3 | 254 | 92 | |
GenBank mRNA | AK149377.1 | - | 2 - 3 | 281 | 92 | |
BC012412.1 | - | 2 - 3 | 254 | 4159892 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |