Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144922.2 | NP_659171.1 | 4 - 5 | 731 | 89 | |
NM_178089.2 | NP_835190.1 | 4 - 5 | 712 | 89 | ||
GenBank mRNA | AK042090.1 | - | 4 - 5 | 712 | 89 | |
AK087545.1 | - | 4 - 5 | 701 | 89 | ||
AK143666.1 | - | 4 - 5 | 774 | 89 | ||
AK155604.1 | - | 4 - 5 | 742 | 89 | ||
AK203240.1 | - | 1 - 2 | 67 | 89 | ||
BC021506.1 | - | 4 - 5 | 755 | 5347099 | 89 | |
BC078642.1 | - | 4 - 5 | 655 | 6329630 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |